Learn More
Zwiebel™ Wood Boxes
Product Code. p-8007696
Change view
Click to view available options
For Use With (Application):
For 1000g Weight
For 100g Weight
For 10g Weight
For 200g Weight
For 50g Weight
Unit Size:
Each
Product Code. For Use With (Application) unitSize
11790513 For 10g Weight Each
11710523 For 50g Weight Each
11720523 For 100g Weight Each
11730523 For 200g Weight Each
11870992 For 1000g Weight Each
5 options
Product Code. 11790513
Brand: Zwiebel™ PBI23853
Only null left
Add to basket
Add to basket
Use to contain E1/E2 classes weights
Empty wooden box
Specifications
Product Type Wood Box
For Use With (Application) For 10g Weight
Quantity 1 Each