missing translation for 'onlineSavingsMsg'
Learn More

WNK-IN-11, MedChemExpress

Product Code. p-200100731
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487277

Brand: Medchem Express HY112094

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WNK-IN-11 is an allosteric With-No-Lysine (WNK) kinase inhibitor, with an IC50 of 4 nM for WNK1.

WNK-IN-11 is an allosteric With-No-Lysine (WNK) kinase inhibitor, with an IC50 of 4 nM for WNK1.

Specifications

Chemical Name or Material WNK-IN-11
CAS 2123489-30-3
Color White
Purity Grade Notes Research
Molecular Formula C21H21Cl2N5OS
Quantity 5 mg
Solubility Information DMSO : 150 mg/mL (324.39 mM; Need ultrasonic)
SMILES O=C(C1=CC(C2=CSC(NC)=N2)=NC=C1Cl)N3CCN(CC4=CC=C(Cl)C=C4)CC3
Molecular Weight (g/mol) 462.4
Formula Weight 462.4
Percent Purity 99.44%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.