Learn More
WISP1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18166-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16848224
  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
to see stock.
Description
Description
The WISP1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human WISP1. This antibody recognizes human, mouse antigen. The WISP1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
WISP1
Polyclonal
Unconjugated
150 μL
8840
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
433 μg/mL
Rabbit
Primary
Human, Mouse