Learn More
WISP1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18166-1-AP-20UL
This item is not returnable.
View return policy
Description
The WISP1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human WISP1. This antibody recognizes human, mouse antigen. The WISP1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| WISP1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 8840 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 433 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |