Learn More
Tork Wiping Paper Plus W1
Product Code. 30243786 Shop All Tork Products
Change view
Click to view available options
Color:
Blue
White
Unit Size:
1 roll
6 rolls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243786 White 6 rolls
30057816 Blue 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30243786 Supplier Tork Supplier No. 130045
  • ONLINE EXCLUSIVE PRICE
    3290.00 NOK
    3125.84 NOK / 6 rolls
    Save 164.16 NOK
    4% Off
View all promotions
Estimated Shipment: 26-11-2026
The 2 ply multi-purpose Tork Wiping Paper Plus is ideal for mopping up liquids and for hand drying.
  • Wipe dirty hands and lightly soiled surfaces with ease with our Tork Wiping Paper Plus.
  • Mop up spills easily, due to the high absorbency of this wiping paper.
  • Tackle tasks requiring strength and absorbency in no time with this multipurpose design.
  • Swiftly carry and dispose of packaging with Tork Easy Handling™ Plastic Packaging.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wiping Paper
Length (English) 557.74 yd.
Length (Metric) 510 m
Width (English) 9.25 in.
Width (Metric) 23.5 cm
Material Paper
Certifications/Compliance EU Ecolabel, ISO 50001, FSC Chain-Of-Custody, OHSAS 18001, HACCP, EMAS, ISO 9001, ISO 14001, FSC Mix Virgin and Rec.fibre
Color White
Packaging Quantity 1500 Sheets
Ply 2-ply
Certifications/Compliance (Europe) EU Ecolabel, ISO 50001, FSC Chain-Of-Custody, OHSAS 18001, HACCP, EMAS, ISO 9001, ISO 14001, FSC Mix Virgin and Rec.fibre