Learn More
Nordvlies WIPEX-SPEZIAL M-roll, 75 g/sqm, app. 20x38 cm, solvent resistent, white
Brand:  Nordvlies 250420
WIPEX-SPEZIAL M-roll, 75 g/sqm, app. 20x38 cm, solvent resistent, white
Product Code. 15710977
  • ONLINE EXCLUSIVE PRICE
    1450.00 NOK
    1170.88 NOK / 6 rolls
    Save 279.12 NOK
    19% Off
Only null left
Estimated Shipment: 18-02-2026