Learn More
Nordvlies WIPEX fleece wipes SOFT, 85 g/sqm, app. 30x38 cm,z-folded, solvent resistent, white
Brand:  Nordvlies 1189
WIPEX fleece wipes SOFT, 85 g/sqm, app. 30x38 cm,z-folded, solvent resistent, white
Product Code. 15700927
  • ONLINE EXCLUSIVE PRICE
    1405.00 NOK
    1135.07 NOK / Pack of 900
    Save 269.93 NOK
    19% Off
Only null left
Estimated Shipment: 28-01-2026