Learn More
Nordvlies WIPEX-CLEANZIE wipes, 50 g/sqm, app. 30x38 cm, z-folded, solvent resistent, white
Brand:  Nordvlies 110335
WIPEX-CLEANZIE wipes, 50 g/sqm, app. 30x38 cm, z-folded, solvent resistent, white
Product Code. 15790907
  • ONLINE EXCLUSIVE PRICE
    945.00 NOK
    643.05 NOK / Pack of 500
    Save 301.95 NOK
    31% Off
Only null left
Estimated Shipment: 22-01-2026