Learn More
Nordvlies WIPEX-BLUETECH Industrial Paper Wipes
Product Code. 15770967
Change view
Click to view available options
Dimensions (L x W):
22 x 36 cm
38 x 36 cm
38 x 38 cm
Unit Size:
1 roll
2 rolls
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15770967 38 x 36 cm 1 roll
15750967 38 x 38 cm 2 rolls
15700977 22 x 36 cm 2 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15770967 Supplier Nordvlies Supplier No. 248007
  • ONLINE EXCLUSIVE PRICE
    686.00 NOK
    517.72 NOK / 1 rolls
    Save 168.28 NOK
    24% Off
View all promotions
Estimated Shipment: 24-11-2026
Blue cleaning paper roll made from selected recycled raw materials, embossed in volume and glued in layers.
  • High absorption capacity of liquid substances, such as oil and water
  • High-performance paper
  • Nordic Swan Ecolabel and EU Ecolabel
  • Industrial, automotive manufacturing, food processing
Specifications
Product Type Heavy-duty Wiper
Length (Metric) 38 cm
Width (Metric) 36 cm
Material Recycled Paper
Autoclavable Not Autoclavable
Color Blue
Disposable Single-use
Dimensions (L x W) 38 x 36 cm
Packaging Quantity 1 Roll of 500 Wipes
Ply 3-ply
Sterility Non-sterile
Thickness (Metric) 3 x 20 g/m2
Green Features Nordic Swan and EU Eco label