Learn More
Berkshire International™ Gamma Wipe™ 67
BIOPHARMA
Product Code. 15270084
Change view
Click to view available options
Dimensions (L x W):
30 x 30 cm
Unit Size:
Pack of 300
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15270084 30 x 30 cm Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15270084 Supplier Berkshire International™ Supplier No. GW67ST60
  • ONLINE EXCLUSIVE PRICE
    6279.00 NOK
    4116.58 NOK / Pack of 300
    Save 2162.42 NOK
    34% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-12-2026
Gamma Wipe™67 is a polyester / cellulose blend wiper recommended for ISO Class 5 and above environments, composed of 45% polyester and 55% cellulose. Each package is gamma irradiated to a 10-6 Sterility Assurance.
  • Gamma irradiated and sterile to SAL 10-6
  • ISO Class 5 to 8
  • Lot number on each pack
  • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Smooth, highly absorbent and durable with low extractable levels
  • Economical
  • Chemically compatible with common cleaning and disinfecting solutions including isopropyl alcohol, phenols, bleaches and quarternary ammonium compounds
  • Abrasion resistant
Specifications
Length (Metric) 30 cm
Width (Metric) 30 cm
Material Hydroentangled Polyester Cellulose
Absorbancy 320 mL/m2
Basis Weight 68.0 g/m2
Certifications/Compliance ISO Class 5 to 8; Cleanroom Class 100 to 100,000; EU Grade A/B/C/D; USP >797<; SAL 10-6 per AAMI guidelines;
Dimensions (L x W) 30 x 30 cm
Format Sheets
For Use With (Application) ISO Class 5 and higher sterile environments; cleaning sterile surfaces and equipment during aseptic processing; general wiping in component prep, compounding and wash areas; general wipedowns in sterile gowning rooms; applying and removing cleaning and disinfecting solutions.