Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Tube Washer
Product Code. 15358638 Shop All SP Scienceware Products
Change view
Click to view available options
Length (Metric):
178 mm
203 mm
Diameter (Metric) Outer:
10 mm Tube Diameter, 14 mm B Diameter
5 mm Tube Diameter, 10 mm B Diameter
Unit Size:
Each
Product Code. Length (Metric) Diameter (Metric) Outer unitSize
15358638 203 mm 5 mm Tube Diameter, 10 mm B Diameter Each
15328638 178 mm 10 mm Tube Diameter, 14 mm B Diameter Each
15348638 178 mm 5 mm Tube Diameter, 10 mm B Diameter Each
3 options
Product Code. 15358638
Brand: Bel-Art™ 660000065
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Wiorks with filter flask and inverted sample tube. Bel-Art SP Scienceware™ Wilmad™ Tube Washer with simple design washing unit is the most economical and reliable for single tube cleaning.
  • Each unit only accommodates NMR tube with OD and length as indicated in specifications
  • Requires filter flask (sold separately; 500ml recommended)
  • Inverted sample tube is inserted into the washer and solvent is introduced into the reservoir using a series of wash bottles.
  • Pulls air through the tube to complete the procedure.
Specifications
Length (Metric) 203 mm
Material Borosilicate Glass
Product Type NMR Tube
Diameter (Metric) Outer 5 mm Tube Diameter, 10 mm B Diameter
Diameter (Metric) Inner 24/40 inner joint
Length (English) 8 in.