Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Spinner Bearing
Ensure shimming quality of your glass tubes. Bel-Art SP Scienceware™ Wilmad™ Spinner Bearing will determine which of your NMR tubes are warped and bent.
Brand: Bel-Art™ 66000-0042
Additional Details : Weight : 0.04310kg
Description
- Useful in labs that wash, dry and reuse precision NMR tubes
- Warped tubes will bind in the spinner bearing while good tubes spin freely
- Keep a tube tester beside every NMR spectrometer for easy access
Specifications
Quartz, Supracil, or Precision tubes | |
For tubes 5mm O.D. | |
SB-5-7 |