Learn More
Bel-Art™ SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System
Product Code. 15318608 Shop All SP Scienceware Products
Change view
Click to view available options
Model:
507-OF-7
OF-30
OFV
Type:
NMR sample tube
Septum
Valve
Unit Size:
Each
Pack of 6
Product Code. Model Type unitSize
15318608 OF-30 Septum Pack of 6
15328608 OFV Valve Each
15398858 507-OF-7 NMR sample tube Each
3 options
Product Code. 15318608
Brand: Bel-Art™ 660000037
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for easy injection of chemicals through a gas-tight syringe for air sensitive samples without using a glove box. Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System is comprised of a 5mm end cap, 2-way valve, septum with cap, barrel coupler and a vacuum connector.
  • The Omni-Fit Tube consists of a 507-PP tube topped by a sturdy 2 in. section of medium-walled tubing which supports the valve system
  • Omni-Fit Sample tubes (507-OF-7) sold separately
  • Omni-Fit valve (OFV) is a complete accessory set
  • Additional septum (OF-30) are available in packages of 6
Due to the nature of glass, Extreme Caution should be exercised when using at elevated or reduced pressures as performance of tubes under pressure or vacuum cannot be guaranteed. Adequate safety shielding should always be used when working in these conditions.
Specifications
Type Septum
Diameter (Metric) 5.0 mm
Model OF-30