Learn More
Bel-Art™ Wilmad™ Replacement O-Ring for Spinner
Description
- Low cost alternative to the full VT version for sample changers.
- Weight and susceptibility matched to originals.
- Can be mixed with originals in sample changer operation.
- Limited VT operation around ambient.
- Does not jam at top of upper barrel during insert operation.
- Tachometer strip now with foil protection for improved life span.
Specifications
Specifications
| For Use With (Equipment) | Agilent/Varian Turbines |
| Type | O-ring |
| Diameter (Metric) Inner | 5 mm |
| Material | Viton |
| Model | TURBINE-ORING-RED |