Learn More
Bel-Art™ Wilmad™ Kel-F Positioning Rod Microcell Small Volume NMR Insert
Product Code. 15328578
Change view
Click to view available options
Type:
Positioning Rod
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
15328578 Positioning Rod Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15328578 Supplier Bel-Art™ Supplier No. 660000010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Insert is ideal for experiments that need to center small volume samples in the Rf coil such as magnetic susceptibility measurement.
  • Sample volume can be as little as 18 μl
  • 3 different inserts with various shapes (Spherical Bulb, Capillary Tube and Cylindrical Bulb)
  • Outer tube (Precision Thin Walled) must be ordered separately depending on magnetic field strength.
  • Not recommended for variable temperature experiments.
  • Use our PTFE Needles (#90630) with syringes to fill and clean sample.
Specifications
For Use With (Equipment) 529-B adapter, Vortex Plugs WG-805C and under
Type Positioning Rod
Description Positioning Rod for inserting small volume microcells
Model 529-C
Only use with Wilmad-LabGlass Precision Thin-Walled Tubes