Learn More
Bel-Art™ SP Scienceware™ Wilmad™ 150mL Suprasil Offset Nitrogen Dewar Flask for Bruker
Product Code. 15358668
Change view
Click to view available options
For Use With (Application):
For studies at 77 K
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
15358668 For studies at 77 K Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15358668 Supplier Bel-Art™ Supplier No. 660000087
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for cavities that have limited space between the reservoir and the drive shaft for iris. Bel-Art SP Scienceware™ Wilmad™ 150mL Suprasil Offset Nitrogen Dewar Flasks provide a convenient alternative to VT systems for studies at 77 K, and are silvered around the reservoir.
  • 150mL flask lasts 30 minutes after a filling
  • Compatible with Bruker™ X-band Cavity
Specifications
For Use With (Equipment) Bruker X-band Cavity
For Use With (Application) For studies at 77 K
Includes Dewar Flask
Model WG-853-B-Q
Volume (Metric) 150 mL