Learn More
Thermo Scientific Chemicals Wig-L-Bug™ Accessories
Product Code. 15456505
Change view
Click to view available options
Height (English):
2 in.
1 ft.
2 ft.
Quantity:
100/Pk.
25/Pk.
5/Pk.
Each
Product Type:
Vial
Vial Adapter
Unit Size:
Each
Pack of 100
Pack of 25
Pack of 5
Product Code. Height (English) Quantity Product Type unitSize
15456505 1 ft. 25/Pk. Vial Pack of 25
15446505 1 ft. 5/Pk. Vial Pack of 5
11405280 2 ft. 25/Pk. Vial Pack of 25
11415280 2 in. 100/Pk. Vial Pack of 100
15402007 - Each Vial Adapter Each
5 options
Product Code. 15456505
Brand: Thermo Scientific Chemicals 042628.KM
Request Bulk or Custom Format
For use with Digital Wig-L-Bug Mixing/Grinding Mill
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Polystyrene vial with polyethylene slip-on cap; 1 ft. long x 0.5 in. dia.
  • Polystyrene vial with polyethylene slip-on cap; 2 ft. long x 0.5 in. dia.
  • Adapter for 2 in. long, 0.5 in. diameter vials
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Height (English) 1 ft.
Quantity 25/Pk.
For Use With (Equipment) Digital Wig-L-Bug mixing/grinding mill
Product Type Vial