Description
- Hygienic nylon brushes
- Robust aluminum handle
- Wide brush shape
Specifications
Specifications
| Product Type | Cleaning Brush |
| Handle Material | Aluminum |
| Diameter (Metric) Bristle | 45 mm |
| For Use With (Equipment) | Tubes with 30 to 45mm Diameter |
| Length (Metric) | 2100 mm |