Learn More
Cole-Parmer™ Wide-Range Pressure Relief Valve
Product Code. 11839642
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Product Code. 11839642
Brand: Cole-Parmer™ 0135066
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for use with Cole-Parmer tubing. These valves are ideal for use with liquid applications.
  • Valves with connections of 1/4 in. or 1/2 in. have a diaphragm made of Viton™ and withstand a maximum inlet pressure of 210psig
  • All other valves have a PTFE shaft design and withstand a maximum inlet pressure of 140psig
  • Shaft design prevents emergency shutdown with a three-seal system that will vent liquid to warn of approaching maintenance
  • All valves are recommended for liquid applications
Specifications
Material Polypropylene
Inlet 1/2 in. NPT(F)
Outlet Port 1/2 in. NPT(F)
Maximum Pressure 210 psig
Product Type Wide-range Pressure Relief Valve