Learn More
Hirschmann™ Wide Neck Square Bottle, 10-pack
Product Code. p-7184619
Click to view available options
Capacity (Metric):
20 mL
30 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
1500 mL
Unit Size:
Pack of 10
Product Code. 16427019
Brand: Hirschmann™ 769066525
Only null left
Add to basket
Add to basket
High quality wide neck square bottle made of HDPE with a screw cap made of LDPE is designed to facilitate easier filling and emptying.
  • Wide neck
  • Square
  • HDPE
  • Starting from size 20 mL and up
  • Natural
  • With screw cap made of LDPE
Specifications
Bottle Surface Transparent
Capacity (Metric) 20 mL
Cleaning Procedure Gas/Chemical/Radiation
Color Natural
Diameter (Metric) Outer Neck 28 mm
Diameter (Metric) Inner Neck 21.5 mm
Material HDPE
Neck ID (Metric) 21.5 mm
No. per Pack 10
Sterility Non-sterile
Thread Size (Metric) GL 28
Closure Color Natural
Content And Storage Store at 5 to 40°C
Closure Included Yes
Shape Square
Height (Metric) 38 mm
Closure Material LDPE
Closure Size GL 28
Diameter (Metric) 35 x 35 mm
Closure Type Screw Cap