Learn More
Hirschmann™ Wide Neck LDPE Bottle, 10-pack
Product Code. 16486979
Change view
Click to view available options
Capacity (Metric):
20 mL
30 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
1500 mL
Unit Size:
Pack of 10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
16486979 20 mL Pack of 10
16406989 30 mL Pack of 10
16426989 50 mL Pack of 10
16446989 100 mL Pack of 10
16466989 250 mL Pack of 10
16486989 500 mL Pack of 10
16406999 1000 mL Pack of 10
16426999 1500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 16486979 Supplier Hirschmann™ Supplier No. 769036525
Only null left
Add to basket
Add to basket
High quality wide neck bottle with a screw cap made of LDPE is designed to facilitate easier filling and emptying.
  • Wide neck
  • LDPE
  • Starting from size 20 mL and up
  • Natural
  • With screw cap
Specifications
Bottle Surface Transparent
Capacity (Metric) 20 mL
Cleaning Procedure Gas/Chemical/Radiation
Color Natural
Diameter (Metric) Outer Neck 25 mm
Diameter (Metric) Inner Neck 18 mm
Material LDPE
Neck ID (Metric) 18 mm
No. per Pack 10
Sterility Non-sterile
Thread Size (Metric) GL 25
Closure Color Natural
Content And Storage Store at 5 to 40°C
Closure Included Yes
Shape Round
Height (Metric) 53 mm
Closure Material LDPE
Closure Size GL 25
Diameter (Metric) 32 mm
Closure Type Screw Cap