Description
- Manufactured from Boro 3.3 glass
- Wide neck for ease of filling
- Acc. to DIN EN ISO 24450
- Provided in packs of 10
Specifications
Specifications
| Diameter (Metric) Outer Neck | 50 mm |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass (3.3) |
| Height (Metric) | 220 mm |
| Neck Style | Wide |
| No. per Pack | 10 |
| Graduated | Yes |
| Diameter (Metric) Outer | 131 mm |