Learn More
CORNING GOSSELIN™ White Polypropylene Pot with Screw Cap
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
125 mL
500 mL
missing translation for 'unitSize'
Pack of 300
Pack of 90
Description
- Break-resistant polypropylene (PP) construction
- White or ivory
- Screw Cap
- Assembled
Specifications
Specifications
| Material | Polypropylene |
| Capacity (Metric) | 125 mL |
| Closure Type | Screw Cap |
| Color | White |
| No. per Case | 300/Cs. |