Learn More
ABEBA™ White Occupational Shoes
Product Code. 15253715
Change view
Click to view available options
Unit Size:
1 pair
Product Code. unitSize
15253715 1 pair
15233715 1 pair
15263715 1 pair
15223715 1 pair
15243715 1 pair
15300202 1 pair
6 options
Product Code. 15253715
Brand: ABEBA™ 9600/41/42
Only null left
Add to basket
Add to basket
Anti-static occupational surgical clogs are non-slip.
  • Autoclavable at 121&deg C for 15 minutes (with no additives) 
  • White 
  • Antistatic 
  • Washable at 60&deg C 
  • Foldable, adjustable heel strap 
  • SRC slip resistance according to EN ISO 20347: 2012 
  • CE EN ISO 20347: 2012 
Specifications
Color White
Style 9600
Product Type Controlled Environments Shoes