Learn More
Coverguard™ White Coverall 5M20
Description
- CE PPE Cat. III
- Limited protection against chemical liquid splash
- Protects against chemical solid particles in the air
- Anti-static protection
- White
- 55 g/m2 SPP + microporous film fabric
- Central zip with adhesive flap
- 3 pans elasticated hood
- Elasticated: waist, wrist with elastic thumbloop, ankle tightening, slingbar
- Self-adhesive chin-strap
Specifications
Specifications
| Size | Large |
| Gender | Unisex |
| Material | PPE |
| Color | White |
| Ankle Style | Elastic |
| Cuff Style | Thumb Loop |
| Hood Type | Elastic with Self-adhesive Chin Strap |
| Closure Type | Central Zip with Adhesive Flap |
| Sterility | Non-sterile |
| Coating | Microporous Film |
| Show More |