Learn More
3M™ White/Blue Combined SMS Laminate Protective Coveralls
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
Description
- Breathable back panel to help improve air ventilation and reduce heat build-up.
- Knit cuffs provide a soft wrist closure for increased comfort compared to elastic closures.
- Elastic waist and ankles for convenience and freedom of movement.
- Two-way zipper with sealable storm flap for convenience and to help provide additional protection.
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | 2X-Large |
| Gender | Unisex |
| Color | Blue, White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Cuff Style | Knit |
| Neck Style | Hooded |
| Sterility | Non-sterile |
| Protection Class | Category III |
| Show More |