Learn More
Thermo Scientific™ White and Black 384-Well Immuno Plates
BIOPHARMA
Product Code. 10385991
Change view
Click to view available options
Color:
Black
White
Unit Size:
Pack of 30
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10385991 White Pack of 30
10395991 Black Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10385991 Supplier Thermo Scientific™ Supplier No. 460372
  • ONLINE EXCLUSIVE PRICE
    4890.00 NOK
    4503.83 NOK / Pack of 30
    Save 386.17 NOK
    7% Off
View all promotions
In Stock
Ajouter au panier
Edge
Ajouter au panier
Bulk Quote Request
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Conduct quantitative and qualitative immunoassays with these white and black 384-well immuno plates.
Conduct quantitative and qualitative immunoassays with Thermo Scientific™ White and Black 384-Well Immuno Plates. Ideal for ELISA and binding assays, these plates are compatible with common instruments. They offer lot-to-lot reliability and well-to-well reproducibility for optimal results.
Features:
  • Plate has SBS footprint, fits standard equipment and is available with flat-bottom wells
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
  • Plate is offered as black or white for optimal choice of application type
  • Certified binding offers homogeneity/reproducibility.
White:
  • Maximum reflection/minimum autofluorescence and autoluminescence
Black:
  • Minimizes crosstalk, back-scattered light and background in fluorescence reading
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering information:
Availability may vary by country
TRUSTED_SUSTAINABILITY
Spécification
Binding Property Hydrophilic
Format Standard
Lid Without Lid
Detection Method Chemiluminescence
Array 16 x 24
Packaging 30 Cases
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 100 μL
Sterility Non-sterile
Surface Treatment MaxiSorp
Well Shape Square, Flat Bottom (F-well)
For Use With (Application) Fluorescence and Luminescence
No. per Case 30
No. per Pack 10
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Well Design F384
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.