Learn More
Thermo Scientific™ White 96-Well Immuno Plates
GREENER_CHOICE
Product Code. 11952984
Click to view available options
Well Shape:
Round, Flat Bottom (F-well)
Surface Treatment:
MaxiSorp
Microfluor 1
Microfluor 2
Microlite 1+
Microlite 2+
PolySorp
Universal Binding
Volume (Metric) Working:
280 μL
330 μL
350 μL
Unit Size:
Pack of 50
Pack of 80
Product Code. 11952984
Brand: Thermo Scientific™ 7905
Only null left
Add to basket
Add to basket
Conduct quantitative and qualitative immunoassays like ELISA and binding assays with these white, 96-well immuno plates.
Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.
Features
  • Plate has SBS footprint — fits standard equipment
  • Available with flat- or round-bottom wells
  • Choice of three surface types for optimal binding
  • Certified binding homogeneity/reproducibility
Choice of Surface:
  • Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
  • Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules
White:
  • Gives maximum reflection
  • Minimum autofluorescence and autoluminescence
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Slightly Hydrophilic
Format Standard
Lid With Lid
Detection Method Chemiluminescence
Array 8 x 12
Packaging 50 Cases
Binding Type Adsorption
Color White
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 2280 μL
No. of Wells 96
Volume (Metric) Working 280 μL
Surface Treatment Microfluor 2
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Fluorescence
No. per Case 50
No. per Pack 10
Plate Blocking None
Target Molecule Antibodies (Glycoproteins) and Proteins