Learn More
Whirl-Pak™ Write-On Stand-Up Thio Sample Bag
GREENER_CHOICE
Product Code. 11787954 Shop All Whirl-Pak Products
Change view
Click to view available options
Thickness (English):
2.5 mil
3 mil
Length (English):
7.25 in.
9 in.
Capacity (English):
4 oz.
10 oz.
Unit Size:
Pack of 100
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (English) Length (English) Capacity (English) unitSize
11787954 3 mil 9 in. 10 oz. Pack of 100
11757954 2.5 mil 7.25 in. 4 oz. Pack of 100
30281137 2.5 mil 7.25 in. 4 oz. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11787954 Supplier Whirl-Pak™ Supplier No. B01403WA
Only null left
Add to basket
Add to basket
Bag stands up completely by itself and contains 10 mg of active sodium thiosulfate to neutralize chlorine in water samples.
  • A special gusset on the bottom of the bag flattens out when water is put in the bag, allowing it to stand up completely by itself.
  • No rack or other holder is needed to keep the bag upright.
  • Forget washing and sterilizing glass bottles and disposing of expensive vials.
  • Sterile and unbreakable
Specifications
Autoclavable Not Autoclavable
Closure Type Tape and Wire
Color Clear
Disposable Single-use
Material Polyethylene
Sterility Sterile
Fill Volume 10 oz.
Latex Free Yes
No. per Pack 100/Pk.
Outer Packaging Cardboard
Protective Inner Packaging Plastic
Quantity 100/Pk.
Sterilization Method Ethylene Oxide Gas
Temperature Range (English) -320°F to +180°F
Temperature Range (Metric) -196°C to +82°C
Thickness (English) 3 mil
Dimensions (L x W) 23 x 11.5 cm (9 x 4.5 in.)
Length (English) 9 in.
Width (English) 4.5 in.
Product Type Liquid Sample Bag
Capacity (English) 10 oz.
Capacity (Metric) 300 mL
Length (Metric) 23 cm
Width (Metric) 11.5 cm
Thickness (Metric) 0.076 mm