Learn More
Whirl-Pak™ Write-On Stand-Up Thio Sample Bag
GREENER_CHOICE
Product Code. 11757954 Shop All Whirl-Pak Products
Change view
Click to view available options
Thickness (English):
2.5 mil
3 mil
Length (English):
7.25 in.
9 in.
Capacity (English):
4 oz.
10 oz.
Unit Size:
Pack of 100
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (English) Length (English) Capacity (English) unitSize
11757954 2.5 mil 7.25 in. 4 oz. Pack of 100
30281137 2.5 mil 7.25 in. 4 oz. Pack of 1000
11787954 3 mil 9 in. 10 oz. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11757954 Supplier Whirl-Pak™ Supplier No. B01402WA
  • ONLINE EXCLUSIVE PRICE
    940.21 NOK
    434.65 NOK / Pack of 100
    Save 505.56 NOK
    53% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Bag stands up completely by itself and contains 10 mg of active sodium thiosulfate to neutralize chlorine in water samples.
  • A special gusset on the bottom of the bag flattens out when water is put in the bag, allowing it to stand up completely by itself.
  • No rack or other holder is needed to keep the bag upright.
  • Forget washing and sterilizing glass bottles and disposing of expensive vials.
  • Sterile and unbreakable
Specifications
Autoclavable Not Autoclavable
Closure Type Tape and Wire
Color Clear with White Block
Disposable Single-use
Material Polyethylene
Sterility Sterile
Fill Volume 4 oz.
Latex Free Yes
No. per Pack 100/Pk.
Outer Packaging Cardboard
Protective Inner Packaging Plastic
Quantity 100/Pk.
Sterilization Method Ethylene Oxide Gas
Temperature Range (English) -320°F to +180°F
Temperature Range (Metric) -196°C to +82°C
Thickness (English) 2.5 mil
Dimensions (L x W) 18.5 x 7.5 cm (7.25 x 3 in.)
Length (English) 7.25 in.
Width (English) 3 in.
Product Type Liquid Sample Bag
Capacity (English) 4 oz.
Capacity (Metric) 118 mL
Length (Metric) 18.5 cm
Width (Metric) 7.5 cm
Thickness (Metric) 0.064 mm