Description
Convenient sterile absorbent sponge sampler in a sterile Whirl-Pak bag
- Designed for testing work surfaces and equipment for Salmonella, Listeria, and other foodborne pathogens
- Dry Speci-Sponge measures 1.5 x 3 x 0.63 in. (3.8 x 7.6 x 1.6 cm); not pretreated with bactericide
- Simply add diluent, wipe test surface, and return sponge to its 18 oz. (532mL) bag
- Self-contained sample can then be shipped for analysis
- Sponge will wipe a large area; bag's puncture-proof tabs reduce damage to sample
- White write-on area allows easy labeling and identification
Order Info
Available with or without sterile glove.
Specifications
Specifications
| Description | With Dry Sponge |
| Autoclavable | Not Autoclavable |
| Capacity (English) | 55 oz. |
| Capacity (Metric) | 1.6 L |
| Closure Type | Tape and Wire |
| Color | Clear |
| Disposable | Single-use |
| For Use With (Application) | For environmental surfaces |
| Includes | Dry Sponge |
| Material | Polyethylene |
| Show More |