Learn More
Nasco Whirl-Pak™ Speci-Sponge™ Environmental Surface Sampling Bags
GREENER_CHOICE
Product Code. 16881590 Shop All Whirl-Pak Products
Click to view available options
Capacity (English):
18 oz.
55 oz.
Includes:
Dry Sponge
Dry Sponge and Glove
Unit Size:
Pack of 100
Product Code. 16881590
Brand: Nasco B01392WA
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Convenient sterile absorbent sponge sampler in a sterile Whirl-Pak bag.
Convenient sterile absorbent sponge sampler in a sterile Whirl-Pak bag
  • Designed for testing work surfaces and equipment for Salmonella, Listeria, and other foodborne pathogens
  • Dry Speci-Sponge measures 1.5 x 3 x 0.63 in. (3.8 x 7.6 x 1.6 cm); not pretreated with bactericide
  • Simply add diluent, wipe test surface, and return sponge to its 18 oz. (532mL) bag
  • Self-contained sample can then be shipped for analysis
  • Sponge will wipe a large area; bag's puncture-proof tabs reduce damage to sample
  • White write-on area allows easy labeling and identification
Order Info
Available with or without sterile glove.
Specifications
Description With Dry Sponge and Glove
Autoclavable Not Autoclavable
Capacity (English) 18 oz.
Capacity (Metric) 532 mL
Closure Type Tape and Wire
Color Clear with White Block
Disposable Single-use
For Use With (Application) For environmental surfaces
Includes Dry Sponge and Glove
Material Polyethylene
Sterility Sterile
Thickness (Metric) 0.038 mm
Fill Volume 18 oz.
No. per Pack 100/Pk.
Outer Packaging Cardboard
Protective Inner Packaging Plastic
Sterilization Method Ethylene Oxide Gas
Temperature Range (English) -320°F to +180°F
Temperature Range (Metric) -196°C to +82°C
Thickness (English) 1.50 mil
Dimensions (L x W) 23 x 11.5 cm (9 x 4 in.)
Length (English) 9 in.
Length (Metric) 23 cm
Width (English) 4 in.
Width (Metric) 11.5 cm
Product Type Sample Bag with Sampling Sponge