Learn More
Whirl-Pak™ Polyethylene Bags
GREENER_CHOICE
Product Code. 11781806 Shop All Whirl-Pak Products
Change view
Click to view available options
Length (Metric):
115 mm
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11781806 115 mm Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11781806 Supplier Whirl Pak Supplier No. B01418WA
Only null left
Add to basket
Add to basket
Whirl-Pak black bag 532mL 115x230mm sterile 500pcs 115x230mm 18oz FDA approved material for light sensitive samples with write-on strip
Specifications
Color Black
For Use With (Application) Light-sensitive samples
Sterility Sterile
Length (Metric) 115 mm