Learn More
WHI-P97, MedChemExpress
Product Code. 16487107
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487107
Brand: Medchem Express HY11067
MedChemExpress WHI-P97 is a potent and selective JAK-3 inhibitor. WHI-P97 is effective in preventing the development allergic asthma in vivo.
WHI-P97 is a potent and selective JAK-3 inhibitor. WHI-P97 is effective in preventing the development allergic asthma in vivo.
Specifications
Chemical Name or Material WHI-P97
CAS 211555-05-4
Color Gray
Purity Grade Notes Research
Molecular Formula C16H13Br2N3O3
Quantity 5 mg
Solubility Information DMSO : 5.88 mg/mL (12.92 mM; Need ultrasonic)
SMILES BrC1=CC(NC2=NC=NC3=CC(OC)=C(OC)C=C32)=CC(Br)=C1O
Molecular Weight (g/mol) 455.1
Formula Weight 455.1
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.