Description
- Wheaton-33™ borosilicate glass
Order Info
Accepts stoppers 06-406-11B and 06-406-12 for freeze drying applications
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 22 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 5 mL |
| For Use With (Application) | For lyophilization and freeze drying applications |
| Thread Style | Screw |
| Volume (English) | 1.4 dr. |
| Closure Type | Screw Cap |
| Certifications/Compliance | ASTM Type I, Class A, USP Type I |
| Height (Metric) | 45 mm |