Learn More
DWK Life Sciences Wheaton™ V Vial™ with Solid-Top Screw Cap, Clear
Shop All Wheaton Science Products ProductsDescription
Includes
14B rubber-lined black phenolic screw caps faced with PTFE
Clear vials are manufactured from Wheaton-33™ low extractable borosilicate glass
Specifications
Specifications
| Material (Liner) | PTFE/14B Rubber |
| Color | Clear |
| Diameter (Metric) Outer | 13 mm |
| Material | Borosilicate Glass (Low Extractable) |
| Volume (Metric) | 1 mL |
| For Use With (Application) | Ideal for small scale reactions, centrifugation, storage, packaging and shipping |
| Graduated | No |
| Style | Conical V |
| Thread Style | Screw |
| Volume (English) | 0.3 dr. |
| Show More |