Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Open-Top Seals
Open-top seal exposes stopper for sample retrieval with a syringe
Brand: DWK Life Sciences 224177-06
1828.75 NOK valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- Unlined
- Closure Type: Seal
Specifications
Red | |
1000/Cs. | |
13 mm |
Aluminum | |
Open Top Cap |