Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Open-Top Seals
GREENER_CHOICE
Product Code. 11708779 Shop All Wheaton Science Products Products
missing translation for 'orderingAttributeHoverText'
Color:
Blue
Green
Natural
Red
Closure Size:
11 mm
13 mm
20 mm
missing translation for 'unitSize'
Pack of 1000
Product Code. 11708779
missing translation for 'mfr': DWK Life Sciences 22417807
Only null left
Add to basket
Add to basket
Open-top seal exposes stopper for sample retrieval with a syringe
  • Unlined
  • Closure Type: Seal
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Material Aluminum
Quantity 1000/Cs.
Type Open Top Cap
Closure Size 20 mm