Learn More
DWK Life Sciences Wheaton™ Uni-Dose Seal Crimper
Compatible with Crimpmaster™ crimping station
Brand:  DWK Life Sciences 224316
Additional Details : Weight : 0.90720kg
Product Code. 11521175
  • 7575.00 NOK / Each
Estimated Shipment: 27-08-2024
Description
  • Ergonomic handle
  • Polished crimping jaws
  • Short handle
  • Spring assembly
  • Adjustable stop
  • Attach 28.5 Uni-Dose seals with this seal crimper