Learn More
DWK Life Sciences Wheaton™ Tissue Grinder Kit
Complete selection of micro tissue grinders packed in a protective carrying case
Brand: DWK Life Sciences 358204
8640.25 NOK valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 1.36080kg
Includes: 1mL Tenbroeck; 0.5mL grinder with 13-425 GPI cap; 0.1 and 0.2mL standard; 2mL Potter-Elvehjem; 1mL tapered
Description
Autoclavable
Order Info
Replacement parts available separately
Specifications
Micro tissue grinder kit | |
1mL Tenbroeck, 0.5mL grinder with 13 to 425mm GPI cap, 0.1 and 0.2mL standard, 2mL Potter-Elvehjem, 1mL tapered |