Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers
Product Code. 11541105 Shop All Wheaton Science Products Products
Click to view available options
Product Type:
2-Leg Lyophilization Stopper
Straight Plug Stopper
Material:
Butyl Rubber
Chlorobutyl Isoprene Blend
Rubber
Unit Size:
Pack of 1000
Product Code. 11541105
Brand: DWK Life Sciences W224100093
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provide airtight covers
  • Flange-type, straight plug stoppers are secured with aluminum crimp seals (06-406-11B)
  • Three-prong plug stoppers are for freeze-dry applications (06-447E, 06-451-56)
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 7 mm
Diameter (Metric) 13 mm
For Use With (Application) Lyophilization
For Use With (Equipment) Fits 7mm I.D., 13mm O.D. mouth of serum bottles and vials
Hardness 50 Durometer
Autoclavable Autoclavable
Color Gray
Material Butyl Rubber
Product Type 2-Leg Lyophilization Stopper