Learn More
DWK Life Sciences Wheaton™ Stainless Steel Support Screens
Product Code. p-3303932 Shop All Wheaton Science Products Products
Change view
Click to view available options
For Use With (Equipment):
25mm Vacuum Filtration Assembly
47mm Vacuum Filtration Assembly
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11591315 47mm Vacuum Filtration Assembly Each
11768959 25mm Vacuum Filtration Assembly Each
2 options
Product Code. 11591315
Brand: DWK Life Sciences 419364
Only null left
Add to basket
Add to basket
For vacuum filtration assembly. Wheaton™ Stainless Steel Support Screens are manufactured from the highest quality materials.
  • Stainless Steel
  • For use with 47mm assembly
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 47mm Vacuum Filtration Assembly
Material Stainless Steel