Learn More
DWK Life Sciences Wheaton™ Snap Plug Caps
Product Code. p-76388 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Closure Size:
8 mm
12 mm
15 mm
Unit Size:
Pack of 1000
Product Code. Color Closure Size unitSize
11591245 Natural 12 mm Pack of 1000
12428892 Green 8 mm Pack of 1000
12458892 Natural 15 mm Pack of 1000
12498882 Natural 8 mm Pack of 1000
12418892 Blue 8 mm Pack of 1000
5 options
Product Code. 11591245
Brand: DWK Life Sciences 242810
Only null left
Add to basket
Add to basket
Manufactured from Polyethylene
  • Closure Style: Snap Cap
  • Liner Material: TFE
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE/Silicone
Color Natural
Material LDPE
Closure Size 12 mm