Learn More
DWK Life Sciences WHEATON™ Septa for Open-Top Caps
Product Code. 11768849 Shop All Wheaton Science Products Products
Change view
Click to view available options
Material:
PTFE-faced Red Rubber
PTFE-faced Silicone
Closure Size:
15 mm
20 mm
22 mm
33 mm
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Closure Size Unit Size
11768849 PTFE-faced Silicone 33 mm Pack of 100
10297123 PTFE-faced Red Rubber 20 mm Pack of 100
11734239 PTFE-faced Red Rubber 15 mm Pack of 100
11531235 PTFE-faced Silicone 22 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11768849 Supplier DWK Life Sciences Supplier No. W240590
  • ONLINE EXCLUSIVE PRICE
    6886.00 NOK
    5320.39 NOK / Pack of 100
    Save 1565.61 NOK
    22% Off
 Restricted Item
Estimated Shipment: 13-11-2026
WHEATON™ Septa for Open-Top Caps are autoclavable septa designed for use with open-top screw caps. Available in a range of sizes and materials including PTFE-faced silicone, natural rubber, and natural red rubber to suit a variety of laboratory sealing needs.
  • Designed for use with open-top screw caps
  • Autoclavable for repeated use
  • Available in multiple sizes to fit a range of cap diameters
  • Available in a variety of materials including PTFE-faced silicone, PTFE-faced natural rubber, and PTFE-faced natural red rubber
Specifications
Color Natural
Material PTFE-faced Silicone
Sterility Non-sterile
Closure Size 33 mm
Diameter (Metric) 33 mm
Autoclavable Autoclavable
Shape Round
Product Type Septa