Learn More
DWK Life Sciences Wheaton™ Septa for Open-Top Caps
Can be used with screw caps
Brand: DWK Life Sciences W240587
4047.00 NOK valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 0.45360kg
Description
- For caps with open tops
- Autoclavable
Specifications
PTFE, Silicone | |
PTFE-faced silicone | |
Septa |
Sterile | |
Autoclavable | |
22 mm |