Learn More
DWK Life Sciences Wheaton™ BioStir™ Magnetic Stirrers
Shop All Wheaton Science Products Products
Click to view available options
Electrical Requirements:
230VAC
Unit Size:
Each
Description
- Programmable
- 5 to 200 RPM
- Remote control and alarm capable
- Waterproof stainless steel housing
- Operating temperature range of 15°C to 40°C
Specifications
Specifications
| Speed | Slow |
| Electrical Requirements | 230VAC |
| Plug Type | Central European |
| Type | Micro-stir |