Learn More
DWK Life Sciences™ Wheaton™ Replacement O-Rings for Dry-Seal Vacuum Desiccator
Shop All Wheaton Science Products Products
Click to view available options
Diameter (Metric) Outer:
100 mm
Unit Size:
Each
Description
- Constructed of slicone rubber
Specifications
Specifications
| Diameter (Metric) Outer | 100 mm |
| Product Type | Replacement O-ring |
| Material | Silicone Rubber |
| For Use With (Application) | Createing an airtight seal when vacuum is applied to dessicator |
| For Use With (Equipment) | For dry-seal vacuum desiccator |
| Diameter (English) Outer | 4 in. |