missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Red Polypropylene Caps for Narrow-Mouth Containers

Product Code. 12493851 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
13-425
20-410
24-410
Unit Size:
Pack of 72
This item is not returnable. View return policy

Product Code. 12493851

Brand: DWK Life Sciences 23950603

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Wheaton leak-resistant narrow mouth bottles

  • Leak-proof design
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 20-410
Certifications/Compliance ISO 9001
For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.