Learn More
DWK Life Sciences Wheaton™ Preassembled Polypropylene Caps
Product Code. 12458862 Shop All Wheaton Science Products Products
Change view
Click to view available options
For Use With (Equipment):
12 mm x 32 mm E-Z Vials
12 mm x 32 mm Vials
15 x 45 mm Vials
Closure Size:
10-425
13-425
8-425
Material (Liner):
PTFE/Silicone
PTFE/Silicone/PTFE
TFE/Silicone
TFE/Styrene-Butadiene Rubber
Unit Size:
Pack of 100
Product Code. For Use With (Equipment) Closure Size Material (Liner) unitSize
12458862 12 mm x 32 mm E-Z Vials 10-425 TFE/Styrene-Butadiene Rubber Pack of 100
12468862 12 mm x 32 mm E-Z Vials 10-425 PTFE/Silicone/PTFE Pack of 100
12478722 12 mm x 32 mm Vials 8-425 PTFE/Silicone Pack of 100
11581245 15 x 45 mm Vials 13-425 PTFE/Silicone Pack of 100
12478862 12 mm x 32 mm E-Z Vials 10-425 TFE/Silicone Pack of 100
5 options
Product Code. 12458862
Brand: DWK Life Sciences 242760
Only null left
Add to basket
Add to basket
Conveniently preassembled with septa
  • Screw Caps
  • Polypropylene
  • Black
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) TFE/Styrene-Butadiene Rubber
Color Black
Material Polypropylene
Closure Size 10-425
For Use With (Equipment) 12 mm x 32 mm E-Z Vials
Product Type Screw Cap