Learn More
DWK Life Sciences Wheaton™ PP Made Delivery Nozzles
Polypropylene Nozzle and Glass Sinker - nozzle connects to the pump tubing for dispensing of liquids
Brand: DWK Life Sciences 374324
555.75 NOK valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.47630kg
Description
- Glass sinker is used to secure tubing when drawing from a container
Specifications
Delivery Nozzle | |
Autoclavable | |
Wheaton peristaltic pumps, dispensing pumps and tubing |
2 and 3 mm | |
Polypropylene |